Dysprosium(III) oxide ≥99.99% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Scientific Chemicals
11318-14
11318-22
11318-36
CAAA11318-14EA
320.74
CAD
CAAA11318-14
CAAA11318-22
CAAA11318-36
Dysprosium(III) oxide ≥99.99% (REO, rare earth oxide basis), REacton®
Dysprosium (III) oxide
MDL: MFCD00010986
Purity: 99.99%
CAS No.: 1308-87-8
Molecular Formula: Dy2O3
Formula Weight: 373.00
Physical Form: Powder
Density: 7.81
Melting Point: 2340°
Merck Reference: 14,3482
MDL No.: MFCD00010986
Formula:
Dy₂O₃ Melting Pt: 2340 °C Density: 7.81 Storage Temperature: Ambient |
MDL Number:
MFCD00010986 CAS Number: 1308-87-8 EINECS: 215-164-0 Merck Index: 14,3482 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...