Dysprosium trichloride hydrate ≥99.99% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Dysprosium(III) chloride hydrate

11317-06 11317-14 11317-22
CAAA11317-06EA 69.05 CAD
CAAA11317-06 CAAA11317-14 CAAA11317-22
Dysprosium trichloride hydrate ≥99.99% (REO, rare earth oxide basis)
Dysprosium trichloride hydrate

MDL: MFCD00010984


Purity: 99.99% Molecular Formula: DyCl3.xH2O Formula Weight: 268.86(anhy) Physical Form: Wet Crystalline Aggregates Sensitivity: Hygroscopic

Formula: Cl₃Dy·xH₂O
MW: 268.86 g/mol (anhy)
Melting Pt: 163 °C
Storage Temperature: Ambient
MDL Number: MFCD00010984
CAS Number: 15059-52-6
EINECS: 233-039-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR