Dysprosium trichloride hydrate ≥99.9% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals
Warning

11316-14 11316-22
CAAA11316-14EA 119.13 CAD
CAAA11316-14 CAAA11316-22
Dysprosium trichloride hydrate ≥99.9% (REO, rare earth oxide basis)
Dysprosium trichloride hydrate

MDL: MFCD00010984 Hydrate level varies due to the nature of this product.


Purity: 99.9% Molecular Formula: DyCl3.xH2O Formula Weight: 268.86(anhy) Physical Form: Crystalline Aggregates Sensitivity: Hygroscopic

Formula: Cl₃Dy·xH₂O
Melting Pt: 163 °C
Storage Temperature: Ambient
MDL Number: MFCD00010984
CAS Number: 15059-52-6
EINECS: 233-039-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR