Dysprosium trinitrate pentahydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Scientific Chemicals
|
Danger
|
11315-14
11315-22
CAAA11315-14EA
195.92
CAD
CAAA11315-14
CAAA11315-22
Dysprosium trinitrate pentahydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Dysprosium trinitrate pentahydrate
MDL: MFCD00149683
Purity: 99.99%
CAS No.: 10031-49-9
Molecular Formula: Dy(NO3)3.5H2O
Formula Weight: 438.59 (348.51anhy)
Physical Form: Crystalline Aggregates
Sensitivity: Hygroscopic
Appearance: Cream to pale yellow
Odor: Odorless
pH: 3
Melting Point: 88.6°
Merck Reference: 14,3482
UN No.: 1477
MDL No.: MFCD00149683
Formula:
DyN₃O₉·5H₂O Melting Pt: 88.6 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149683 CAS Number: 10031-49-9 EINECS: 233-410-5 UN: 1477 ADR: 5.1,III Merck Index: 14,3482 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...