Erbium trifluoride, anhydrous ≥99.99% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals
Warning

11312-09 11312-18
CAAA11312-09EA 199.08 CAD
CAAA11312-09 CAAA11312-18
Erbium trifluoride, anhydrous ≥99.99% (REO, rare earth oxide basis), REacton®
Erbium trifluoride

MDL: MFCD00016074


Purity: 99.99% CAS No.: 13760-83-3 Molecular Formula: ErF3 Formula Weight: 224.28 Physical Form: Granular/Powder Density: 6.4 Melting Point: 1350° MDL No.: MFCD00016074

Formula: ErF₃
Boiling Pt: 2200 °C (1013 hPa)
Melting Pt: 1350 °C
Density: 6.4
Storage Temperature: Ambient
MDL Number: MFCD00016074
CAS Number: 13760-83-3
EINECS: 237-356-3
UN: 3288
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR