Erbium(III) oxide ≥99.99% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Dierbium trioxide, Erbium oxide

11309-06 11309-14 11309-22 11309-36
CAAA11309-06EA 111.86 CAD
CAAA11309-06 CAAA11309-14 CAAA11309-22 CAAA11309-36
Erbium(III) oxide ≥99.99% (REO, rare earth oxide basis), REacton®
Erbium (III) oxide

MDL: MFCD00010991


Purity: 99.99% CAS No.: 12061-16-4 Molecular Formula: Er2O3 Formula Weight: 382.56 Physical Form: 20 Micron Powder Density: 8.64 Melting Point: 2400° Merck Reference: 14,3646 MDL No.: MFCD00010991

Formula: Er₂O₃
MW: 382.52 g/mol
Melting Pt: 2420
Density: 8.64
Storage Temperature: Ambient
MDL Number: MFCD00010991
CAS Number: 12061-16-4
EINECS: 235-045-7
Merck Index: 14,3646

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR