Erbium trinitrate pentahydrate ≥99.99% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals
Danger

11306-14 11306-22
CAAA11306-14EA 290.72 CAD
CAAA11306-14 CAAA11306-22
Erbium trinitrate pentahydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Erbium trinitrate pentahydrate

MDL: MFCD00149691


Purity: 99.99% CAS No.: 10031-51-3 Molecular Formula: Er(NO3)3.5H2O Formula Weight: 443.35 (353.27anhy) Physical Form: Crystalline Aggregates Sensitivity: Hygroscopic UN No.: 1477 MDL No.: MFCD00149691

Formula: ErN₃O₉·5H₂O
Melting Pt: 130 °C (-4H₂O)
Storage Temperature: Ambient
MDL Number: MFCD00149691
CAS Number: 10031-51-3
EINECS: 233-436-7
UN: 1477
ADR: 5.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR