Erbium trichloride hydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Scientific Chemicals
|
Warning
|
11305-14
11305-22
CAAA11305-14EA
169.06
CAD
CAAA11305-14
CAAA11305-22
Erbium trichloride hydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Erbium trichloride hydrate
MDL: MFCD00010989
Purity: 99.99%
CAS No.: 19423-85-9
Molecular Formula: ErCl3.xH2O
Formula Weight: 273.62(anhy)
Physical Form: Crystalline Aggregates
Sensitivity: Hygroscopic
MDL No.: MFCD00010989
Formula:
Cl₃Er·6H₂O Melting Pt: 168 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010989 CAS Number: 10025-75-9 EINECS: 233-385-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...