Erbium trichloride hydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Scientific Chemicals
|
Warning
|
11304-14
11304-22
CAAA11304-14EA
115.5
CAD
CAAA11304-14
CAAA11304-22
Erbium trichloride hydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Erbium trichloride hydrate
MDL: MFCD00010989
Purity: 99.9%
CAS No.: 19423-85-9
Molecular Formula: ErCl3.xH2O
Formula Weight: 273.62(anhy)
Physical Form: Crystalline Aggregates
Sensitivity: Hygroscopic
MDL No.: MFCD00010989
Formula:
Cl₃Er·H₂O MW: 291.63 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00010989 CAS Number: 19423-85-9 EINECS: 233-385-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...