Europium(III) fluoride, anhydrous ≥99.98% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals
Warning

11302-04 11302-06 11302-14
CAAA11302-04EA 175.38 CAD
CAAA11302-04 CAAA11302-06 CAAA11302-14
Europium(III) fluoride, anhydrous ≥99.98% (REO, rare earth oxide basis), REacton®
Europium (III) fluoride

MDL: MFCD00010998


Purity: 99.98% CAS No.: 13765-25-8 Molecular Formula: EuF3 Formula Weight: 208.96 Physical Form: Powder Sensitivity: Hygroscopic Boiling Point: 2280° MDL No.: MFCD00010998

Formula: EuF₃
MW: 208.96 g/mol
Boiling Pt: >2280 °C
Melting Pt: 1390 °C
Storage Temperature: Ambient
MDL Number: MFCD00010998
CAS Number: 13765-25-8
EINECS: 237-368-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR