Europium(III) oxide ≥99.9% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Scientific Chemicals
11300-06
11300-14
11300-22
CAAA11300-06EA
206.98
CAD
CAAA11300-06
CAAA11300-14
CAAA11300-22
Europium(III) oxide ≥99.9% (REO, rare earth oxide basis), REacton®
Europium (III) oxide
MDL: MFCD00010997
Purity: 99.9%
CAS No.: 1308-96-9
Molecular Formula: Eu2O3
Formula Weight: 351.92
Physical Form: Powder
Sensitivity: Hygroscopic
Merck Reference: 14,3904
MDL No.: MFCD00010997
Formula:
Eu₂O₃ Melting Pt: 2291 °C Density: 7.42 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010997 CAS Number: 1308-96-9 EINECS: 215-165-6 Merck Index: 14,3904 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...