Europium(III) chloride hydrate ≥99.9% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals

11298-06 11298-14
CAAA11298-06EA 169.06 CAD
CAAA11298-06 CAAA11298-14
Europium(III) chloride hydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Europium(III) chloride hydrate

MDL: MFCD00010994


Purity: 99.9% Molecular Formula: EuCl3.xH2O Formula Weight: 258.32(anhy) Physical Form: Crystalline Aggregates Sensitivity: Hygroscopic Odor: Odorless Merck Reference: 14,3904 MDL No.: MFCD00010994

Formula: Cl₃Eu·xH₂O
Melting Pt: 158 °C
Density: 4.89 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00217047
CAS Number: 13759-92-7
EINECS: 233-040-4
Merck Index: 14,3904

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR