Europium trinitrate hydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Europium (III) nitrate hydrate
11296-06
11296-14
CAAA11296-06EA
292.3
CAD
CAAA11296-06
CAAA11296-14
Europium trinitrate hydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Europium trinitrate hydrate
MDL: MFCD00149700
Purity: 99.99%
CAS No.: 100587-95-9
Molecular Formula: Eu(NO3)3.xH2O (x≈6)
Formula Weight: 337.97(anhy)
Physical Form: Crystalline Aggregates
Appearance: Colorless to pink
Melting Point: 85°
Merck Reference: 14,3904
UN No.: 1477
MDL No.: MFCD00149700
Formula:
EuN₃O₉·H₂O MW: 337.98 g/mol (anhy) Melting Pt: 85 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149700 CAS Number: 100587-95-9 EINECS: 233-380-3 UN: 1477 ADR: 5.1,III Merck Index: 14,3904 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...