Digadolinium trioxide ≥99.99% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Gadolinium (III) oxide

11290-09 11290-18 11290-30 11290-A1
CAAA11290-09EA 74.89 CAD
CAAA11290-09 CAAA11290-18 CAAA11290-30 CAAA11290-A1
Digadolinium trioxide ≥99.99% (REO, rare earth oxide basis), REacton®
Digadolinium trioxide

MDL: MFCD00011026


Purity: 99.99% CAS No.: 12064-62-9 Molecular Formula: Gd2O3 Formula Weight: 362.50 Physical Form: 10 Micron Powder Density: 7.407 Melting Point: 2330° Merck Reference: 14,4326 MDL No.: MFCD00011026

Formula: Gd₂O₃
MW: 362.5 g/mol
Melting Pt: 2330 °C
Density: 7.407
Storage Temperature: Ambient
MDL Number: MFCD00011026
CAS Number: 12064-62-9
EINECS: 235-060-9
Merck Index: 14,4326

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR