Digadolinium trioxide ≥99.999% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Gadolinium (III) oxide

11289-14 11289-22 11289-36
CAAA11289-14EA 189.6 CAD
CAAA11289-14 CAAA11289-22 CAAA11289-36
Digadolinium trioxide ≥99.999% (REO, rare earth oxide basis), REacton®
Digadolinium trioxide

MDL: MFCD00011026


Purity: 99.999% CAS No.: 12064-62-9 Molecular Formula: Gd2O3 Formula Weight: 362.50 Physical Form: 10 Micron Powder Density: 7.407 Melting Point: 2330° Merck Reference: 14,4326 MDL No.: MFCD00011026

Formula: Gd₂O₃
MW: 362.5 g/mol
Melting Pt: 2410
Density: 7.407
Storage Temperature: Ambient
MDL Number: MFCD00011026
CAS Number: 12064-62-9
EINECS: 235-060-9
Merck Index: 14,4326

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR