Gadolinium trichloride hexahydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Scientific Chemicals
|
Warning
|
11286-06
11286-14
11286-22
11286-A1
CAAA11286-06EA
58.93
CAD
CAAA11286-06
CAAA11286-14
CAAA11286-22
CAAA11286-A1
Gadolinium trichloride hexahydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Gadolinium trichloride hexahydrate
MDL: MFCD00149732
Purity: 99.9%
CAS No.: 13450-84-5
Molecular Formula: GdCl3.6H2O
Formula Weight: 371.70 (263.61anhy)
Physical Form: Crystalline Aggregates
Density: 2.424
Appearance: White
Odor: Characteristic
Merck Reference: 14,4326
MDL No.: MFCD00149732
Formula:
Cl₃Gd·6H₂O Density: 2.424 Storage Temperature: Ambient |
MDL Number:
MFCD00149732 CAS Number: 13450-84-5 EINECS: 233-386-6 Merck Index: 14,4326 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...