Holmium trifluoride, anhydrous ≥99.99% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Holmium (III) fluoride

11278-06 11278-14
CAAA11278-06EA 143.94 CAD
CAAA11278-06 CAAA11278-14
Holmium trifluoride, anhydrous ≥99.99% (REO, rare earth oxide basis), REacton®
Holmium trifluoride

MDL: MFCD00016142


Purity: 99.99% CAS No.: 13760-78-6 Molecular Formula: HoF3 Formula Weight: 221.93 Physical Form: Crystalline Lump Density: 7.6 Melting Point: 1143° UN No.: 3288 MDL No.: MFCD00016142

Formula: HoF₃
MW: 221.93 g/mol
Melting Pt: 1143 °C
Density: 7.6
Storage Temperature: Ambient
MDL Number: MFCD00016142
CAS Number: 13760-78-6
EINECS: 237-352-1
UN: 3288
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR