Holmium trichloride hexahydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Scientific Chemicals
|
Warning
|
11277-14
11277-22
CAAA11277-14EA
232.26
CAD
CAAA11277-14
CAAA11277-22
Holmium trichloride hexahydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Holmium trichloride hexahydrate
MDL: MFCD00149739
Purity: 99.9%
CAS No.: 14914-84-2
Molecular Formula: Cl3Ho.6H2O
Formula Weight: 379.38 (271.29anhy)
Storage Temperature: 2°C to 8°C
Physical Form: Crystalline Aggregates
Sensitivity: Hygroscopic
Odor: Odorless
Melting Point: 718°
Merck Reference: 14,4727
MDL No.: MFCD00149739
Formula:
Cl₃Ho·6H₂O Melting Pt: 718 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00149739 CAS Number: 14914-84-2 EINECS: 233-387-1 Merck Index: 14,4727 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...