Lanthanum(III) fluoride, anhydrous ≥99.99% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Scientific Chemicals
|
Warning
|
11273-18
11273-30
CAAA11273-18EA
194.34
CAD
CAAA11273-18
CAAA11273-30
Lanthanum(III) fluoride, anhydrous ≥99.99% (REO, rare earth oxide basis), REacton®
Lanthanum (III) fluoride
MDL: MFCD00011073
Purity: 99.99%
CAS No.: 13709-38-1
Molecular Formula: F3La
Formula Weight: 195.90
Physical Form: Crystalline
Density: 5.936
Melting Point: 1493°
MDL No.: MFCD00011073
Formula:
F₃La Boiling Pt: ∼2327 °C (1013 hPa) Melting Pt: 1493 °C Density: 5.936 Storage Temperature: Ambient |
MDL Number:
MFCD00011073 CAS Number: 13709-38-1 EINECS: 237-252-8 UN: 3288 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...