Lanthanum(III) oxide ≥99.9% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals
Warning

11272-22 11272-36
CAAA11272-22EA 136.99 CAD
CAAA11272-22 CAAA11272-36
Lanthanum(III) oxide ≥99.9% (REO, rare earth oxide basis), REacton®
Lanthanum (III) oxide

MDL: MFCD00011071


Purity: 99.9% CAS No.: 1312-81-8 Molecular Formula: La2O3 Formula Weight: 325.82 Physical Form: Powder Density: 6.51 Melting Point: 2307° Boiling Point: 4200° Merck Reference: 14,5363 MDL No.: MFCD00011071

Formula: La₂O₃
MW: 325.81 g/mol
Boiling Pt: 4200
Melting Pt: 2315
Density: 6.51
Storage Temperature: Ambient
MDL Number: MFCD00011071
CAS Number: 1312-81-8
EINECS: 215-200-5
Merck Index: 14,5363

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR