Lanthanum, compound with nickel (1:5) ≥99.9% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Scientific Chemicals
11271-18
11271-90
CAAA11271-18EA
170.64
CAD
CAAA11271-18
CAAA11271-90
Lanthanum, compound with nickel (1:5) ≥99.9% (REO, rare earth oxide basis), REacton®
Lanthanum, compound with nickel (1:5)
Lanthanum nickel act as a superconducting intermetallic phase in lanthanum nickel boro-nitride.
Purity: 99.9%
CAS No.: 12196-72-4
Molecular Formula: LaNi5
Physical Form: Pieces, Intermetallic, Packaged under argon
Sensitivity: Air and Moisture Sensitive
Odor: Odorless
UN No.: 3178
MDL No.: MFCD00198177
Formula:
LaNi₅ |
MDL Number:
MFCD00198177 CAS Number: 12196-72-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...