Lanthanum(III) nitrate hexahydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Scientific Chemicals
|
Danger
|
11267-06
11267-18
11267-30
11267-A1
CAAA11267-06EA
61.78
CAD
CAAA11267-06
CAAA11267-18
CAAA11267-30
CAAA11267-A1
Lanthanum(III) nitrate hexahydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Lanthanum (III) nitrate hexahydrate
MDL: MFCD00149751
Purity: 99.99%
CAS No.: 10277-43-7
Molecular Formula: La(NO3)3.6H2O
Formula Weight: 433.01 (324.92anhy)
Physical Form: Crystalline Aggregates
Odor: Odorless
Merck Reference: 14,5363
UN No.: 1477
MDL No.: MFCD00149751
Formula:
La(NO₃)₃·6H₂O Boiling Pt: 126 °C (1013 hPa) Melting Pt: 40 °C Density: 2.347 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149751 CAS Number: 10277-43-7 EINECS: 233-238-0 UN: 1477 ADR: 5.1,III Merck Index: 14,5363 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...