Lutetium trichloride hexahydrate ≥99.9% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals

11260-03 11260-06
CAAA11260-03EA 141.57 CAD
CAAA11260-03 CAAA11260-06
Lutetium trichloride hexahydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Lutetium trichloride hexahydrate

MDL: MFCD00011099


Purity: 99.9% CAS No.: 15230-79-2 Molecular Formula: LuCl3.6H2O Formula Weight: 389.42 (281.33anhy) Physical Form: Crystalline Aggregates

Formula: Cl₃Lu·6H₂O
Storage Temperature: Ambient
MDL Number: MFCD00011099
CAS Number: 15230-79-2
EINECS: 233-240-1
Merck Index: 14,5615

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR