Lutetium(III) oxide ≥99.99% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Lutetium oxide, Dilutetium trioxide

11256-03 11256-06 11256-14
CAAA11256-03EA 184.86 CAD
CAAA11256-03 CAAA11256-06 CAAA11256-14
Lutetium(III) oxide ≥99.99% (REO, rare earth oxide basis), REacton®
Lutetium (III) oxide

MDL: MFCD00011100


Purity: 99.99% CAS No.: 12032-20-1 Molecular Formula: Lu2O3 Formula Weight: 397.94 Physical Form: Powder Density: 9.420 Melting Point: 2487° Merck Reference: 14,5615 MDL No.: MFCD00011100

Formula: Lu₂O₃
MW: 397.93 g/mol
Melting Pt: 2510
Density: 9.420
Storage Temperature: Ambient
MDL Number: MFCD00011100
CAS Number: 12032-20-1
EINECS: 234-764-3
Merck Index: 14,5615

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR