Neodymium(III) chloride hydrate ≥99.99% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals
Warning

11252-14 11252-22
CAAA11252-14EA 178.54 CAD
CAAA11252-14 CAAA11252-22
Neodymium(III) chloride hydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Neodymium(III) chloride hydrate

Neodymium(III) chloride hydrate is used for production of neodymium metal. It also finds application in lasers and fiber amplifiers. It serves as dopants for silica based as well as plastic fibers. It plays an important role of additives in organic light emitting diodes resulting in improved LED emission spectrum. In addition, it is used in catalysis of polymerization of various dienes. It is used to modify titanium dioxide for use as a photocatalyst. Its coating on aluminum and its alloy protect them from corrosion. It is also extensively used as fluorescent labels for organic molecules.


Purity: 99.99% CAS No.: 10024-93-8 Molecular Formula: NdCl3.xH2O Formula Weight: 250.60(anhy) Physical Form: Wet Crystalline Aggregates Odor: Odorless Merck Reference: 14,6450 MDL No.: MFCD00011132

Formula: NdCl₃·xH₂O
Melting Pt: 124 °C
Density: 2.28 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011132
CAS Number: 13477-89-9
EINECS: 233-031-5
Merck Index: 14,6450

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR