Neodymium(III) oxide ≥99.99% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Scientific Chemicals
Synonyms:
Dineodymium trioxide, Neodymium(III)-oxide, Neodymium oxide
11250-14
11250-22
11250-A1
CAAA11250-14EA
181.7
CAD
CAAA11250-14
CAAA11250-22
CAAA11250-A1
Neodymium(III) oxide ≥99.99% (REO, rare earth oxide basis), REacton®
Neodymium (III) oxide
MDL: MFCD00011134
Purity: 99.99%
CAS No.: 1313-97-9
Molecular Formula: Nd2O3
Formula Weight: 336.48
Physical Form: Powder
Density: 7.24
Melting Point: 2272°
Merck Reference: 14,6450
MDL No.: MFCD00011134
Formula:
Nd₂O₃ MW: 336.48 g/mol Boiling Pt: 3760 Melting Pt: 2272 °C Density: 7.24 Storage Temperature: Ambient |
MDL Number:
MFCD00011134 CAS Number: 1313-97-9 EINECS: 215-214-1 Merck Index: 14,6450 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...