Neodymium trifluoride, anhydrous ≥99.9% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Scientific Chemicals
|
Warning
|
11246-14
11246-22
CAAA11246-14EA
243.32
CAD
CAAA11246-14
CAAA11246-22
Neodymium trifluoride, anhydrous ≥99.9% (REO, rare earth oxide basis), REacton®
Neodymium trifluoride
MDL: MFCD00011135
Purity: 99.9%
CAS No.: 13709-42-7
Molecular Formula: F3Nd
Formula Weight: 201.24
Physical Form: Crystalline Lump
Density: 6.5
Appearance: Pale purple
Odor: Odorless
Melting Point: 1410°
MDL No.: MFCD00011135
Formula:
F₃Nd MW: 201.24 Boiling Pt: 2327 Melting Pt: 1410 °C Density: 6.5 Storage Temperature: Ambient |
MDL Number:
MFCD00011135 CAS Number: 13709-42-7 EINECS: 237-253-3 UN: 3288 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...