Praseodymium trifluoride, anhydrous ≥99.9895% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals
Danger

11241-09 11241-18
CAAA11241-04EA 142.94 CAD
CAAA11241-04 CAAA11241-09 CAAA11241-18
Praseodymium trifluoride, anhydrous ≥99.9895% (REO, rare earth oxide basis), REacton®
Praseodymium trifluoride

MDL: MFCD00016287


Purity: 99.9895% CAS No.: 13709-46-1 Molecular Formula: F3Pr Formula Weight: 197.90 Physical Form: Powder Density: 6.2 Appearance: Green Odor: Odorless Melting Point: 1395° UN No.: 3288 MDL No.: MFCD00016287

Formula: F₃Pr
MW: 197.9
Melting Pt: 1395
Density: 6.2
Storage Temperature: Ambient
MDL Number: MFCD00016287
CAS Number: 13709-46-1
EINECS: 237-254-9
UN: 3288
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR