Praseodymium(III) nitrate hexahydrate ≥99.99% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals
Danger

11240-14 11240-22
CAAA11240-14EA 278.08 CAD
CAAA11240-14 CAAA11240-22
Praseodymium(III) nitrate hexahydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Praseodymium trinitrate hexahydrate

MDL: MFCD00149826


Purity: 99.99% CAS No.: 15878-77-0 Molecular Formula: Pr(NO3)3.6H2O Formula Weight: 435.01 (326.92anhy) Physical Form: Crystalline Aggregates Sensitivity: Hygroscopic UN No.: 1477 MDL No.: MFCD00149826

Formula: N₃O₉Pr·6H₂O
Melting Pt: 56 °C
Density: 2.233 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149826
CAS Number: 15878-77-0
EINECS: 233-796-5
UN: 1477
ADR: 5.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR