Praseodymium trichloride hydrate ≥99.9% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Praseodymium (III) chloride hydrate

11237-14 11237-22
CAAA11237-14EA 127.35 CAD
CAAA11237-14 CAAA11237-22
Praseodymium trichloride hydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Praseodymium trichloride hydrate

MDL: MFCD00149828 Soluble in water and alcohol


Purity: 99.9% CAS No.: 19423-77-9 Molecular Formula: PrCl3.xH2O Formula Weight: 247.27(anhy) Physical Form: Crystalline Aggregates Odor: Odorless Merck Reference: 14,7712 MDL No.: MFCD00149828

Formula: Cl₃Pr·H₂O
MW: 265.28
Melting Pt: 115 °C
Storage Temperature: Ambient
MDL Number: MFCD00149828
CAS Number: 19423-77-9
EINECS: 233-794-4
Merck Index: 14,7712

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR