Praseodymium(III,IV) oxide ≥99.99% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals
Warning

11235-14 11235-22
CAAA11235-14EA 371.3 CAD
CAAA11235-14 CAAA11235-22
Praseodymium(III,IV) oxide ≥99.99% (REO, rare earth oxide basis), REacton®
Praseodymium (III,IV) oxide

MDL: MFCD00011178


Purity: 99.99% CAS No.: 12037-29-5 Molecular Formula: Pr6O11 Formula Weight: 1021.43 Physical Form: Powder Melting Point: 2500° MDL No.: MFCD00011178

Formula: O₁₁Pr₆
MW: 1021.44 g/mol
Melting Pt: 2500 °C
Storage Temperature: Ambient
MDL Number: MFCD00011178
CAS Number: 12037-29-5
EINECS: 234-857-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR