Samarium trichloride, hydrate ≥99.9% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals
Warning

11231-14 11231-22
CAAA11231-14EA 114.55 CAD
CAAA11231-14 CAAA11231-22
Samarium trichloride, hydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Samarium trichloride

MDL: MFCD00011235


Purity: 99.9% CAS No.: 10361-82-7 Molecular Formula: SmCl3.xH2O Formula Weight: 256.71(anhy) Physical Form: Crystalline Aggregates Sensitivity: Hygroscopic Merck Reference: 14,8347 MDL No.: MFCD00011235

Formula: Cl₃Sm·H₂O
Melting Pt: 110 °C (-5H₂O)
Storage Temperature: Ambient
MDL Number: MFCD00011235
CAS Number: 19423-79-1
EINECS: 233-797-0
Merck Index: 14,8347

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR