Samarium(III) oxide ≥99.9% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals

11229-14 11229-22 11229-36 11229-A1
CAAA11229-14EA 135.25 CAD
CAAA11229-14 CAAA11229-22 CAAA11229-36 CAAA11229-A1
Samarium(III) oxide ≥99.9% (REO, rare earth oxide basis), REacton®
Samarium (III) oxide

MDL: MFCD00011237


Purity: 99.9% CAS No.: 12060-58-1 Molecular Formula: Sm2O3 Formula Weight: 348.70 Physical Form: Powder Refractive Index: 1.97 Melting Point: 2300° Merck Reference: 14,8347 MDL No.: MFCD00011237

Formula: O₃Sm₂
MW: 348.72 g/mol
Melting Pt: 2325
Density: 7.68 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011237
CAS Number: 12060-58-1
EINECS: 235-043-6
Merck Index: 14,08347

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR