Samarium(III) fluoride, anhydrous ≥99.99% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Scientific Chemicals
|
Danger
|
11226-04
11226-09
11226-18
CAAA11226-04EA
83.42
CAD
CAAA11226-04
CAAA11226-09
CAAA11226-18
Samarium(III) fluoride, anhydrous ≥99.99% (REO, rare earth oxide basis), REacton®
Samarium (III) fluoride
MDL: MFCD00049540
Purity: 99.99%
CAS No.: 13765-24-7
Molecular Formula: SmF3
Formula Weight: 207.35
Physical Form: Powder
Sensitivity: Hygroscopic
UN No.: 3288
MDL No.: MFCD00049540
Formula:
F₃Sm MW: 207.36 Boiling Pt: 2323 Melting Pt: 1306 °C Density: 4.362 g/cm³ (2327 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00049540 CAS Number: 13765-24-7 EINECS: 237-367-3 UN: 3288 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...