Samarium trinitrate hexahydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Scientific Chemicals
|
Danger
|
11224-06
11224-14
11224-22
CAAA11224-06EA
55.3
CAD
CAAA11224-06
CAAA11224-14
CAAA11224-22
Samarium trinitrate hexahydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Samarium trinitrate hexahydrate
MDL: MFCD00149857
Purity: 99.99%
CAS No.: 13759-83-6
Molecular Formula: Sm(NO3)3.xH2O (x≈6)
Formula Weight: 336.36(anhy)
Physical Form: Crystalline Aggregates
Density: 2.375
Appearance: Pale yellow
Odor: Odorless
pH: ca. 3
Melting Point: 78-79°
UN No.: 1477
MDL No.: MFCD00149857
Formula:
N₃O₉Sm·6H₂O Melting Pt: 78…79 °C Density: 2.375 Storage Temperature: Ambient |
MDL Number:
MFCD00149857 CAS Number: 13759-83-6 EINECS: 233-798-6 UN: 1477 ADR: 5.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...