Scandium(III) nitrate hydate ≥99.99% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Scientific Chemicals
|
Danger
|
11221-03
11221-06
CAAA11221-03EA
259.12
CAD
CAAA11221-03
CAAA11221-06
Scandium(III) nitrate hydate ≥99.99% (REO, rare earth oxide basis), REacton®
Scandium (III) nitrate
MDL: MFCD00011220
Purity: 99.99%
CAS No.: 13465-60-6
Molecular Formula: Sc(NO3)3.xH2O
Formula Weight: 230.97(anhy)
Physical Form: Crystalline Aggregates
Sensitivity: Hygroscopic
Merck Reference: 14,8392
UN No.: 1477
MDL No.: MFCD00011220
Formula:
N₃O₉Sc·H₂O Storage Temperature: Ambient |
MDL Number:
MFCD00011220 CAS Number: 107552-14-7 EINECS: 236-701-5 UN: 1477 ADR: 5.1,III Merck Index: 14,8392 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...