Scandium(III) chloride hexahydrate ≥99.99% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals

11219-03 11219-06
CAAA11219-03EA 240.16 CAD
CAAA11219-03 CAAA11219-06
Scandium(III) chloride hexahydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Scandium (III) chloride hexahydrate

MDL: MFCD00149852


Purity: 99.99% CAS No.: 20662-14-0 Molecular Formula: ScCl3.6H2O Formula Weight: 259.41 (151.32anhy) Physical Form: Crystalline Aggregates Sensitivity: Hygroscopic Merck Reference: 14,8392 MDL No.: MFCD00149852

Formula: Cl₃Sc·6H₂O
MW: 259.41 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149852
CAS Number: 20662-14-0
EINECS: 233-799-1
Merck Index: 14,8392

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR