Terbium(III) chloride hexahydrate ≥99.99% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals
Warning

11210-09 11210-18
CAAA11210-09EA 241.74 CAD
CAAA11210-09 CAAA11210-18
Terbium(III) chloride hexahydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Terbium (III) chloride hexahydrate

MDL: MFCD00149870


Purity: 99.99% CAS No.: 13798-24-8 Molecular Formula: TbCl3.6H2O Formula Weight: 373.38 (265.29anhy) Physical Form: Crystalline Aggregates Odor: Odorless Merck Reference: 14,9157 MDL No.: MFCD00149870

Formula: Cl₃Tb·6H₂O
MW: 373.38 g/mol
Melting Pt: 145
Density: 4.35 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149870
CAS Number: 13798-24-8
EINECS: 233-132-4
Merck Index: 14,9157

Order Now

Specification Test Results

Form Crystalline Aggregates


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR