Terbium(III) chloride hexahydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Scientific Chemicals
|
Warning
|
11209-09
11209-18
CAAA11209-09EA
199.08
CAD
CAAA11209-09
CAAA11209-18
Terbium(III) chloride hexahydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Terbium (III) chloride hexahydrate
MDL: MFCD00149870
Very soluble in water
Purity: 99.9%
CAS No.: 13798-24-8
Molecular Formula: TbCl3.6H2O
Formula Weight: 373.38 (265.29anhy)
Physical Form: Crystalline Aggregates
Sensitivity: Hygroscopic
Solubility: Very soluble in water
Merck Reference: 14,9157
MDL No.: MFCD00149870
Formula:
Cl₃Tb·6H₂O MW: 373.38 g/mol Melting Pt: 145 °C Density: 4.35 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149870 CAS Number: 13798-24-8 EINECS: 233-132-4 Merck Index: 14,9157 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...