Terbium(III,IV) oxide ≥99.99% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals

11208-06 11208-14
CAAA11208-06EA 298.62 CAD
CAAA11208-06 CAAA11208-14
Terbium(III,IV) oxide ≥99.99% (REO, rare earth oxide basis), REacton®
Terbium (III,IV) oxide

MDL: MFCD00083151


Purity: 99.99% CAS No.: 12037-01-3 Molecular Formula: Tb4O7 Formula Weight: 747.69 Physical Form: Powder Odor: Odorless Merck Reference: 14,9157 MDL No.: MFCD00083151

Formula: O₇Tb₄
Melting Pt: 2340 °C
Storage Temperature: Ambient
MDL Number: MFCD00083151
CAS Number: 12037-01-3
EINECS: 234-856-3
Merck Index: 14,9157

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR