Terbium(III,IV) oxide ≥99.9% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Scientific Chemicals
11207-09
11207-18
CAAA11207-09EA
328.64
CAD
CAAA11207-09
CAAA11207-18
Terbium(III,IV) oxide ≥99.9% (REO, rare earth oxide basis), REacton®
Terbium (III,IV) oxide
MDL: MFCD00083151
Purity: 99.9%
CAS No.: 12037-01-3
Molecular Formula: Tb4O7
Formula Weight: 747.69
Physical Form: Powder
Odor: Odorless
Merck Reference: 14,9157
MDL No.: MFCD00083151
Formula:
O₇Tb₄ Melting Pt: 2340 °C Storage Temperature: Ambient |
MDL Number:
MFCD00083151 CAS Number: 12037-01-3 EINECS: 234-856-3 Merck Index: 14,9157 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...