Thulium(III) nitrate hydrate ≥99.99% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals
Danger

11203-04 11203-09
CAAA11203-04EA 186.44 CAD
CAAA11203-04 CAAA11203-09 CAAA11203-18
Thulium(III) nitrate hydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Thulium(III) nitrate hydrate

MDL: MFCD00149875


Purity: 99.99% Molecular Formula: Tm(NO3)3.xH2O (x≈5) Formula Weight: 354.95(anhy) Physical Form: Wet Crystalline Aggregates UN No.: 1477 MDL No.: MFCD00149875

Formula: N₃O₉Tm·xH₂O
Storage Temperature: Ambient
MDL Number: MFCD00149875
CAS Number: 36548-87-5
EINECS: 239-069-9
UN: 1477
ADR: 5.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR