Thulium(III) oxide ≥99.99% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Scientific Chemicals
|
Warning
|
11199-04
11199-09
CAAA11199-04EA
276.5
CAD
CAAA11199-04
CAAA11199-09
Thulium(III) oxide ≥99.99% (REO, rare earth oxide basis), REacton®
Thulium (III) oxide
MDL: MFCD00011285
Purity: 99.99%
CAS No.: 12036-44-1
Molecular Formula: O3Tm2
Formula Weight: 385.87
Physical Form: Powder
Appearance: Pale green
Melting Point: 2425°
Merck Reference: 14,9394
MDL No.: MFCD00011285
Formula:
O₃Tm₂ Melting Pt: 2425 °C Density: 8.6 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011285 CAS Number: 12036-44-1 EINECS: 234-851-6 Merck Index: 14,9394 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...