Ytterbium(III) nitrate hydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Ytterbium trinitrate hydrate
11196-09
11196-18
CAAA11196-09EA
176.96
CAD
CAAA11196-09
CAAA11196-18
Ytterbium(III) nitrate hydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Ytterbium(III) nitrate hydrate
MDL: MFCD00149883
Purity: 99.99%
Molecular Formula: Yb(NO3)3.xH2O (x≈5-6)
Formula Weight: 359.05(anhy)
Physical Form: Crystalline Aggregates
Sensitivity: Hygroscopic
Appearance: White
Merck Reference: 14,10105
UN No.: 1477
MDL No.: MFCD00149883
Formula:
Yb(NO₃)₃·xH₂O MW: 359.05 g/mol (anhy) Storage Temperature: Ambient |
MDL Number:
MFCD00149883 CAS Number: 35725-34-9 EINECS: 237-384-6 UN: 1477 ADR: 5.1 III Merck Index: 14,10105 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...