Ytterbium(III) chloride hydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Scientific Chemicals
|
Warning
|
11194-09
11194-18
CAAA11194-09EA
164.32
CAD
CAAA11194-09
CAAA11194-18
Ytterbium(III) chloride hydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Ytterbium (III) chloride hydrate
MDL: MFCD00149885
Purity: 99.99%
CAS No.: 19423-87-1
Molecular Formula: YbCl3.xH2O (x~6)
Formula Weight: 279.40(anhy)
Physical Form: Crystalline Aggregates
Density: 2.58
Appearance: Green
Odor: Odorless
Melting Point: 150-155°
Merck Reference: 14,10105
MDL No.: MFCD00149885
Formula:
Cl₃Yb·H₂O Melting Pt: 150…155 °C Density: 2.58 Storage Temperature: Ambient |
MDL Number:
MFCD00149885 CAS Number: 19423-87-1 EINECS: 233-800-5 Merck Index: 14,10105 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...