Ytterbium(III) chloride hydrate ≥99.9% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals
Warning

11193-09 11193-18
CAAA11193-09EA 86.27 CAD
CAAA11193-09 CAAA11193-18
Ytterbium(III) chloride hydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Ytterbium (III) chloride hydrate

MDL: MFCD00149885


Purity: 99.9% CAS No.: 19423-87-1 Molecular Formula: YbCl3.xH2O (x≈6) Formula Weight: 279.40(anhy) Physical Form: Wet Crystalline Aggregates Density: 2.58 Appearance: Green Odor: Odorless Melting Point: 150-155° Merck Reference: 14,10105 MDL No.: MFCD00149885

Formula: Cl₃Yb·H₂O
MW: 297.41 g/mol
Melting Pt: 150…155 °C
Density: 2.58
Storage Temperature: Ambient
MDL Number: MFCD00149885
CAS Number: 19423-87-1
EINECS: 233-800-5
Merck Index: 14,10105

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR