Ytterbium(III) oxide ≥99.9% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Scientific Chemicals
11191-14
11191-22
11191-A1
CAAA11191-14EA
123.08
CAD
CAAA11191-14
CAAA11191-22
CAAA11191-A1
Ytterbium(III) oxide ≥99.9% (REO, rare earth oxide basis), REacton®
Ytterbium (III) oxide
MDL: MFCD00011290
Purity: 99.9%
CAS No.: 1314-37-0
Molecular Formula: Yb2O3
Formula Weight: 394.08
Physical Form: Powder
Density: 9.2
Melting Point: 2227°
Merck Reference: 14,10105
MDL No.: MFCD00011290
Formula:
O₃Yb₂ Melting Pt: 2227 °C Density: 9.2 Storage Temperature: Ambient |
MDL Number:
MFCD00011290 CAS Number: 1314-37-0 EINECS: 215-234-0 Merck Index: 14,10105 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...