Ytterbium(III) fluoride, anhydrous ≥99.99% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Scientific Chemicals
|
Danger
|
11189-03
11189-09
11189-18
CAAA11189-03EA
63.2
CAD
CAAA11189-03
CAAA11189-09
CAAA11189-18
Ytterbium(III) fluoride, anhydrous ≥99.99% (REO, rare earth oxide basis), REacton®
Ytterbium (III) fluoride
MDL: MFCD00049615
Purity: 99.99%
CAS No.: 13760-80-0
Molecular Formula: YbF3
Formula Weight: 230.04
Physical Form: Lump/Powder
Density: 8.2
Melting Point: 1157°
UN No.: 3288
MDL No.: MFCD00049615
Formula:
F₃Yb Boiling Pt: 2227 °C (1013 hPa) Melting Pt: 1157 °C Density: 8.2 Storage Temperature: Ambient |
MDL Number:
MFCD00049615 CAS Number: 13760-80-0 EINECS: 237-354-2 UN: 3288 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...