L(+)-Ascorbic acid 98+%
Supplier: Thermo Scientific Chemicals
11188-22
11188-36
11188-A3
CAAA11188-22EA
53.74
CAD
CAAA11188-22
CAAA11188-36
CAAA11188-A3
L(+)-Ascorbic acid 98+%
L(+)-Ascorbic acid
MDL: MFCD00064328
Beilstein Registry No.: 84272
Purity: 98+%
CAS No.: 50-81-7
Molecular Formula: C6H8O6
Formula Weight: 176.12
Physical Form: Powder
Density: 1.65
Melting Point: ca 191° dec.
Beilstein Reference: 84272
Merck Reference: 14,830
MDL No.: MFCD00064328
Formula:
C₆H₈O₆ Boiling Pt: 553 °C (1013 hPa) Melting Pt: ∼ 191 °C (dec.) Density: 1.65 Storage Temperature: Ambient |
MDL Number:
MFCD00064328 CAS Number: 50-81-7 EINECS: 200-066-2 REACH: exempt Merck Index: 14,830 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...