Yttrium (III) nitrate hydrate ≥99.99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Yttrium trinitrate hydrate

11187-06 11187-14 11187-22 11187-59
CAAA11187-06EA 70.15 CAD
CAAA11187-06 CAAA11187-14 CAAA11187-22 CAAA11187-59
Yttrium (III) nitrate hydrate ≥99.99%
Yttrium (III) nitrate hydrate

MDL: MFCD00011469


Purity: 99.99% Molecular Formula: Y(NO3)3.xH2O Formula Weight: 274.92(anhy) Physical Form: Crystalline Aggregates Sensitivity: Hygroscopic Merck Reference: 14,10107 MDL No.: MFCD00011469

Formula: N₃O₉Y·H₂O
MW: 274.92 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00011469
CAS Number: 140191-40-8
EINECS: 233-802-6
UN: 1477
ADR: 5.1 III
Merck Index: 14,10107

Order Now

Specification Test Results

Form Crystalline Aggregates


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR